Anti-CRIM1 Mouse monoclonal antibody [clone: 9G343]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7935-66EA 3700 DKK
USBIC7935-66
Anti-CRIM1 Mouse monoclonal antibody [clone: 9G343]
Antistoffer
Recommended dilution: Immunochemistry: 3?g/ml; ELISA: 0. 03ng/ml

Type: Primary
Antigen: CRIM1 (cysteine rich transmembrane BMP regulator 1 (chordin-like))
Clonality: Monoclonal
Clone: 9G343
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR