Anti-CRLK Mouse monoclonal antibody [clone: 12K74]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7925-50GEA 4320 DKK
USBIC7925-50G
Anti-CRLK Mouse monoclonal antibody [clone: 12K74]
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: CRLK
Clonality: Monoclonal
Clone: 12K74
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR