Anti-CRKL Mouse polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7925-01AEA 3500 DKK
USBIC7925-01A
Anti-CRKL Mouse polyclonal antibody
Antistoffer
Type: Primary
Antigen: CRKL (v-crk avian sarcoma virus CT10 oncogene homolog-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR