Anti-CRK Mouse monoclonal antibody [clone: 12K73]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7924-10KEA 4320 DKK
USBIC7924-10K
Anti-CRK Mouse monoclonal antibody [clone: 12K73]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CRK (v-crk avian sarcoma virus CT10 oncogene homolog)
Clonality: Monoclonal
Clone: 12K73
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR