Anti-TDGF1 Mouse Monoclonal Antibody [clone: 9F614]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7922-10JEA 2870 DKK
USBIC7922-10J
Anti-TDGF1 Mouse Monoclonal Antibody [clone: 9F614]
Antistoffer
Recommended dilution: ELISA: 1:8000

Type: Primary
Antigen: TDGF1 (teratocarcinoma-derived growth factor 1)
Clonality: Monoclonal
Clone: 9F614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR