Anti-TDGF1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7922-10B-BIOTEA 4620 DKK
USBIC7922-10B-BIOT
Anti-TDGF1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: TDGF1 (teratocarcinoma-derived growth factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR