Anti-CRIM1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2602]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7921-90-FITCEA 0 DKK
USBIC7921-90-FITC
Anti-CRIM1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2602]
Type: Primary
Antigen: CRIM1 (cysteine rich transmembrane BMP regulator 1 (chordin-like))
Clonality: Monoclonal
Clone: 10B2602
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR