Anti-CRIM1 Mouse monoclonal antibody [clone: 6E4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7921-90AEA 4790 DKK
USBIC7921-90A
Anti-CRIM1 Mouse monoclonal antibody [clone: 6E4]
Antistoffer
Recommended dilution: Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: CRIM1 (cysteine rich transmembrane BMP regulator 1 (chordin-like))
Clonality: Monoclonal
Clone: 60000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR