Anti-CREG1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2601]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7918-70-PEEA 0 DKK
USBIC7918-70-PE
Anti-CREG1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2601]
Type: Primary
Antigen: CREG1 (cellular repressor of E1A-stimulated genes 1)
Clonality: Monoclonal
Clone: 10B2601
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR