Anti-PTGDR2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7916-05GEA 4730 DKK
USBIC7916-05G
Anti-PTGDR2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: PTGDR2 (prostaglandin D2 receptor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR