Anti-CRTAP Mouse monoclonal antibody Biotin [clone: 10B2607]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-96-BIOTEA 0 DKK
USBIC7915-96-BIOT
Anti-CRTAP Mouse monoclonal antibody Biotin [clone: 10B2607]
Type: Primary
Antigen: CRTAP (cartilage associated protein)
Clonality: Monoclonal
Clone: 10B2607
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR