Anti-CREB3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2599]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-41-APEA 0 DKK
USBIC7915-41-AP
Anti-CREB3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2599]
Type: Primary
Antigen: CREB3 (cAMP responsive element binding protein 3)
Clonality: Monoclonal
Clone: 10B2599
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR