Anti-CREB Rabbit monoclonal antibody [clone: 9B146]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-25EA 3850 DKK
USBIC7915-25
Anti-CREB Rabbit monoclonal antibody [clone: 9B146]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:250 ChImmunoprecipitation: 1:50 Immunochemistry: 1:400; Immunofluorescence: 1:1600; Flow Cytometry: 1:200

Type: Primary
Antigen: CREB
Clonality: Monoclonal
Clone: 9B146
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR