Anti-CREB1, p43 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-18EA 5010 DKK
USBIC7915-18
Anti-CREB1, p43 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; SupershImmunofluorescencet: 0.5-1?l

Type: Primary
Antigen: CREB1, p43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR