Anti-CREB1 Mouse monoclonal antibody [clone: 5F2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-17BEA 4790 DKK
USBIC7915-17B
Anti-CREB1 Mouse monoclonal antibody [clone: 5F2]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CREB1 (cAMP responsive element binding protein 1)
Clonality: Monoclonal
Clone: 5F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR