Anti-CREB1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-05PEA 4320 DKK
USBIC7915-05P
Anti-CREB1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CREB1 (cAMP responsive element binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR