Anti-CREB Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-05LEA 2840 DKK
USBIC7915-05L
Anti-CREB Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot:1:500-1:1000; Immunochemistry:1:50-1:100

Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR