Anti-CREB Rabbit Monoclonal Antibody (Alexa Fluor® 488)) [clone: 7H16]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-04CEA 3850 DKK
USBIC7915-04C
Anti-CREB Rabbit Monoclonal Antibody (Alexa Fluor® 488)) [clone: 7H16]
Antistoffer
Recommended dilution: Flow Cytometry 1:10; Immunofluorescence:1:10; Immunochemistry: 1:10
Do not freeze!

Type: Primary
Antigen: CREB
Clonality: Monoclonal
Clone: 7H16
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR