Anti-CREB Mouse monoclonal antibody [clone: 9A121]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-02BEA 4830 DKK
USBIC7915-02B
Anti-CREB Mouse monoclonal antibody [clone: 9A121]
Antistoffer
Type: Primary
Antigen: CREB
Clonality: Monoclonal
Clone: 9A121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR