Anti-CREB Mouse monoclonal antibody [clone: 7H84]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7915-01BEA 4320 DKK
USBIC7915-01B
Anti-CREB Mouse monoclonal antibody [clone: 7H84]
Antistoffer
Recommended dilution: Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: CREB
Clonality: Monoclonal
Clone: 7H84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR