Anti-Creatinine Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7911-46EA 6900 DKK
USBIC7911-46
Anti-Creatinine Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 2?g/ml

Type: Primary
Antigen: Creatinine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR