Anti-Creatine Phosphokinase Sheep polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7910-40AEA 6040 DKK
USBIC7910-40A
Anti-Creatine Phosphokinase Sheep polyclonal antibody
Antistoffer
Type: Primary
Antigen: Creatine Phosphokinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu