Anti-Creatine Kinase Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7910-29-20EA 2680 DKK
USBIC7910-29-100 USBIC7910-29-20
Anti-Creatine Kinase Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: Creatine Kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR