Anti-Creatine Kinase MM Isoenzyme Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7910-28DEA 3160 DKK
USBIC7910-28D
Anti-Creatine Kinase MM Isoenzyme Goat polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Creatine Kinase MM Isoenzyme
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR