Anti-Creatine Kinase BB Isoenzyme Mouse monoclonal antibody [clone: 10B2596]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7910-14GEA 0 DKK
USBIC7910-14G
Anti-Creatine Kinase BB Isoenzyme Mouse monoclonal antibody [clone: 10B2596]
Type: Primary
Antigen: Creatine Kinase BB Isoenzyme
Clonality: Monoclonal
Clone: 10B2596
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR