Anti-CKB Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7910-13DEA 3460 DKK
USBIC7910-13D
Anti-CKB Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:2000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: CKB (creatine kinase, brain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR