Anti-Cre Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7909-02EA 4180 DKK
USBIC7909-02
Anti-Cre Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Cre
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR