Anti-CPEB1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7908-01CEA 3460 DKK
USBIC7908-01C
Anti-CPEB1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: CPEB1 (cytoplasmic polyadenylation element binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR