Anti-CRP Rat monoclonal antibody [clone: 10K78]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-15FEA 5320 DKK
USBIC7907-15F
Anti-CRP Rat monoclonal antibody [clone: 10K78]
Antistoffer
Recommended dilution: ELISA:2-8?g/ml

Type: Primary
Antigen: CRP (C-reactive protein, pentraxin-related)
Clonality: Monoclonal
Clone: 10K78
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR