Anti-C-Reactive Protein Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-15B-100EA 5140 DKK
USBIC7907-15B-100
Anti-C-Reactive Protein Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:5000

Type: Primary
Antigen: C-Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR