Anti-C-Reactive Protein Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-14AEA 3290 DKK
USBIC7907-14A
Anti-C-Reactive Protein Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: C-Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR