Anti-C-Reactive Protein Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-12EEA 2800 DKK
USBIC7907-12E
Anti-C-Reactive Protein Goat polyclonal antibody
Type: Primary
Antigen: C-Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR