Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 10K173]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-10AEA 3290 DKK
USBIC7907-10A
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 10K173]
Antistoffer
Recommended dilution: ELISA: 1 ?g/ml

Type: Primary
Antigen: C-Reactive Protein
Clonality: Monoclonal
Clone: 10K173
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR