Anti-C-Reactive Protein Sheep polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7907-04L2EA 3580 DKK
USBIC7907-04L2
Anti-C-Reactive Protein Sheep polyclonal antibody
Antistoffer
Type: Primary
Antigen: C-Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human