Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 12K202]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-03UEA 3870 DKK
USBIC7907-03U
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 12K202]
Antistoffer
Type: Primary
Antigen: C-Reactive Protein
Clonality: Monoclonal
Clone: 12K202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR