Anti-C-Reactive Protein Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7907-03JEA 2400 DKK
USBIC7907-03J
Anti-C-Reactive Protein Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 3?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: C-Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR