Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 9F422]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7907-01ZEA 3580 DKK
USBIC7907-01Z
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 9F422]
Antistoffer
Type: Primary
Antigen: C-Reactive Protein
Clonality: Monoclonal
Clone: 9F422
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestil nu