Anti-CRABP2 Mouse Monoclonal Antibody [clone: AT2E11]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7906-71HEA 4730 DKK
USBIC7906-71H
Anti-CRABP2 Mouse Monoclonal Antibody [clone: AT2E11]
Antistoffer
Recommended dilution: Western Blot: 1:250-1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: Monoclonal
Clone: AT2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR