Anti-CRABP2 Mouse Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7906-71FEA 3500 DKK
USBIC7906-71F
Anti-CRABP2 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR