Anti-CRABP2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7906-71EEA 5850 DKK
USBIC7906-71E
Anti-CRABP2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR