Anti-Cellular Retonic Acid Binding Protein 1 Mouse monoclonal antibody [clone: 10D216]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7906-70DEA 2800 DKK
USBIC7906-70D
Anti-Cellular Retonic Acid Binding Protein 1 Mouse monoclonal antibody [clone: 10D216]
Antistoffer
Recommended dilution: Western Blot: 1:500-1000

Type: Primary
Antigen: Cellular Retonic Acid Binding Protein 1
Clonality: Monoclonal
Clone: 10D216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR