Anti-CRAT Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7906-16BEA 3700 DKK
USBIC7906-16B
Anti-CRAT Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CRAT (carnitine O-acetyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR