Anti-CPSF3 Mouse monoclonal antibody [clone: 10B2590]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7906-05DEA 0 DKK
USBIC7906-05D
Anti-CPSF3 Mouse monoclonal antibody [clone: 10B2590]
Type: Primary
Antigen: CPSF3 (cleavage and polyadenylation specific factor 3, 73kDa)
Clonality: Monoclonal
Clone: 10B2590
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR