Anti-C-Peptide I Mouse Monoclonal Antibody [clone: 10B356]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7905-18ZEA 3970 DKK
USBIC7905-18Z
Anti-C-Peptide I Mouse Monoclonal Antibody [clone: 10B356]
Antistoffer
Type: Primary
Antigen: C-Peptide I
Clonality: Monoclonal
Clone: 10B356
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR