Anti-C-Peptide I Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7905-18XEA 4620 DKK
USBIC7905-18X
Anti-C-Peptide I Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:500

Type: Primary
Antigen: C-Peptide I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR