Anti-C-Peptide Mouse Monoclonal Antibody [clone: 9E33]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7905-10AEA 3920 DKK
USBIC7905-10A
Anti-C-Peptide Mouse Monoclonal Antibody [clone: 9E33]
Antistoffer
Type: Primary
Antigen: C-Peptide
Clonality: Monoclonal
Clone: 9E+33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR