Anti-C-Peptide Mouse Monoclonal Antibody [clone: 9L127]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7905-09BEA 4100 DKK
USBIC7905-09B
Anti-C-Peptide Mouse Monoclonal Antibody [clone: 9L127]
Antistoffer
Type: Primary
Antigen: C-Peptide
Clonality: Monoclonal
Clone: 9L127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human