Anti-C Peptide Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7905-02AEA 3680 DKK
USBIC7905-02A
Anti-C Peptide Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Radio Immunoassay: 1:2000; Western Blot: 1:100; Immunochemistry: 1:100

Type: Primary
Antigen: C Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR