Anti-C-Peptide Mouse Monoclonal Antibody [clone: 10g77]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7905-01TEA 3310 DKK
USBIC7905-01T
Anti-C-Peptide Mouse Monoclonal Antibody [clone: 10g77]
Antistoffer
Recommended dilution: ELISA: 1-2?g/ml; Immunochemistry: 20-40 ?g/ml

Type: Primary
Antigen: C-Peptide
Clonality: Monoclonal
Clone: 10g77
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR