Anti-COX4I1, COX4I2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89XEA 3460 DKK
USBIC7904-89X
Anti-COX4I1, COX4I2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:4000; Western Blot: 0. 01-0. 03?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: COX4I1, COX4I2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR